Please login to your online account to display your discounted pricing

2,6-Dichloroindophenol, Sodium Salt ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloroindophenol, Sodium Salt
CAS 620-45-1
Purity Grade ACS Grade
Molecular Formula O:C6H2Cl2:NC6H4ONa
Formula Weight 290.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK305410

 
avantor performance
305454
Glass 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 290.08
Quantity 10g
Molecular Formula O:C6H2Cl2:NC6H4ONa
CAS 620-45-1
Packaging Glass
Chemical Name or Material 2, 6-Dichloroindophenol, Sodium Salt