missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloroindophenol, Sodium Derivative, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$549.40 - $848.65

Chemical Identifiers

CAS 620-45-1
Molecular Formula O:C6H2(Cl2):NC6H4ONa
Formula Weight 290.08
Synonym 2,6-Dichlorophenalindephenol Sodium Salt
Chemical Name or Material 2,6-Dichloroindophenol, Sodium Derivative
Grade Baker Analyzed™ ACS Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02002070 Avantor Performance
H11601
Wide Mouth Amber Glass Bottle 5g Each for $549.40
02002071 Avantor Performance
H11602
Wide Mouth Amber Glass Bottle 10g Each for $848.65
Specifications Description & Specifications

Chemical Identifiers

620-45-1
290.08
2,6-Dichloroindophenol, Sodium Derivative
O:C6H2(Cl2):NC6H4ONa
2,6-Dichlorophenalindephenol Sodium Salt
Baker Analyzed™ ACS Reagent

Specifications

5g
O:C6H2(Cl2):NC6H4ONa
Wide Mouth Amber Glass Bottle
2,6-Dichloroindophenol, Sodium Derivative
620-45-1
290.08
2,6-Dichlorophenalindephenol Sodium Salt
Baker Analyzed™ ACS Reagent