Please login to your online account to display your discounted pricing

2,6-Dichloroindophenol, Sodium Derivative, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 10g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  H11602

 View more versions of this product

Catalog No. 02002071

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 290.08
Chemical Name or Material 2,6-Dichloroindophenol, Sodium Derivative
Molecular Formula O:C6H2(Cl2):NC6H4ONa
Packaging Wide Mouth Amber Glass Bottle
CAS 620-45-1
Synonym 2,6-Dichlorophenalindephenol Sodium Salt
Quantity 10g

Specifications

Chemical Name or Material 2,6-Dichloroindophenol, Sodium Derivative
CAS 620-45-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula O:C6H2(Cl2):NC6H4ONa
Formula Weight 290.08
Synonym 2,6-Dichlorophenalindephenol Sodium Salt