missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorodiphenylamine, 98%

Manufacturer:  Alfa Aesar™ H5584803

 View more versions of this product

Catalog No. AAH5584803



Specifications

Specifications

2,6-Dichlorodiphenylamine
Brown
1g
C12H9Cl2N
#
C1=CC=C(C=C1)NC2=C(C=CC=C2Cl)Cl
238.111
238.12
98%
110°C (230°F)
50°C to 53°C
15307-93-4
MFCD00269648
HDUUZPLYVVQTKN-UHFFFAOYSA-N
2,6-dichloro-N-phenylaniline
84867
Solid

Chemical Identifiers

15307-93-4
238.111
HDUUZPLYVVQTKN-UHFFFAOYSA-N
84867
C1=CC=C(C=C1)NC2=C(C=CC=C2Cl)Cl
C12H9Cl2N
MFCD00269648
#
2,6-dichloro-N-phenylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 239-349-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents