Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorocinnamic acid, predominantly trans, 98%

$55.89 - $171.77

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorocinnamic acid
CAS 5345-89-1
Name Note predominantly trans
Molecular Formula C9H6Cl2O2
Formula Weight 217.05
MDL Number MFCD00004374
Beilstein 1953654
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0561414

  SDS
alfa aesar™
L0561414
25g Each for $171.77

AAL0561406

  SDS
alfa aesar™
L0561406
5g Each for $55.89
Description & Specifications

Specifications

Formula Weight 217.05
Chemical Name or Material 2,6-Dichlorocinnamic acid
MDL Number MFCD00004374
CAS 5345-89-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note predominantly trans
Assay 98%
Molecular Formula C9H6Cl2O2
Beilstein 1953654
Melting Point 193° to 197°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number UD3333800
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.