Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorobenzylideneacetone, 98%

$75.11 - $75.11

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorobenzylideneacetone
CAS 81559-89-9
Molecular Formula C10H8Cl2O
Formula Weight 215.08
MDL Number MFCD00051609
Synonym 4-(2,6-Dichlorophenyl)-3-buten-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0570606

 
alfa aesar™
L0570606
5g Each for $75.11
Description & Specifications

Specifications

Formula Weight 215.08
Assay 98%
Molecular Formula C10H8Cl2O
MDL Number MFCD00051609
CAS 81559-89-9
Boiling Point 135° to 136°C (2mmHg)
Synonym 4-(2,6-Dichlorophenyl)-3-buten-2-one
Quantity 5g
Chemical Name or Material 2,6-Dichlorobenzylideneacetone