Please login to your online account to display your discounted pricing

Alfa Aesar™ (2,6-Dichlorobenzyl)triphenylphosphonium chloride, 98%

$86.66 - $1,057.49

Chemical Identifiers

Chemical Name or Material (2,6-Dichlorobenzyl)triphenylphosphonium chloride
CAS 18583-54-5
Molecular Formula C25H20Cl3P
Formula Weight 457.78
MDL Number MFCD00061392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1765106

  SDS
alfa aesar™
A1765106
5g Each for $86.66

AAA1765122

  SDS
alfa aesar™
A1765122
100g Each for $1,057.49
Description & Specifications

Specifications

Formula Weight 457.78
Chemical Name or Material (2,6-Dichlorobenzyl)triphenylphosphonium chloride
Assay 98%
Molecular Formula C25H20Cl3P
MDL Number MFCD00061392
CAS 18583-54-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.