Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorobenzyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorobenzyl chloride
CAS 2014-83-7
Molecular Formula C7H5Cl3
Formula Weight 195.48
MDL Number MFCD00000897
UN Number UN3261
Beilstein 472282
Synonym alpha,2,6-Trichlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1331014

 
alfa aesar™
A1331014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1331030

 
alfa aesar™
A1331030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1331022

 
alfa aesar™
A1331022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.48
Chemical Name or Material 2,6-Dichlorobenzyl chloride
Odor Pungent
Assay 97%
Molecular Formula C7H5Cl3
MDL Number MFCD00000897
CAS 2014-83-7
UN Number UN3261
Boiling Point 117° to 119°C (14mmHg)
Beilstein 472282
Melting Point 36° to 39°C
Flash Point >110°C (230°F)
Synonym alpha,2,6-Trichlorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.