missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorobenzyl bromide, 97%

$62.15 - $189.68

Chemical Identifiers

CAS 20443-98-5
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00000577
InChI Key PDFGFQUSSYSWNI-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
PubChem CID 30159
IUPAC Name 2-(bromomethyl)-1,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)CBr)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0171014 View Documents Alfa Aesar™
L0171014
25g Each for $62.15
 
AAL0171022 View Documents Alfa Aesar™
L0171022
100g Each for $189.68
 
Specifications

Chemical Identifiers

20443-98-5
239.921
PDFGFQUSSYSWNI-UHFFFAOYSA-N
30159
C1=CC(=C(C(=C1)Cl)CBr)Cl
C7H5BrCl2
MFCD00000577
2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
2-(bromomethyl)-1,3-dichlorobenzene

Specifications

>110°C (230°F)
54°C to 57°C
20443-98-5
MFCD00000577
637351
PDFGFQUSSYSWNI-UHFFFAOYSA-N
2-(bromomethyl)-1,3-dichlorobenzene
30159
97%
Pungent
25g
C7H5BrCl2
UN3261
2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
C1=CC(=C(C(=C1)Cl)CBr)Cl
239.921
239.93
2,6-Dichlorobenzyl bromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : XS7966000