Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorobenzyl alcohol, 99%

$195.46 - $195.46

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorobenzyl alcohol
CAS 15258-73-8
Molecular Formula C7H6Cl2O
Formula Weight 177.03g/mol
MDL Number MFCD00004609
Beilstein 1938356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1219618

 
alfa aesar™
A12196-18
50g Each for $195.46
Description & Specifications

Specifications

Boiling Point 150°C (25mmHg)
Melting Point 95° to 99°C
Quantity 50g
Chemical Name or Material 2,6-Dichlorobenzyl alcohol
CAS 15258-73-8
Assay 99%
Molecular Formula C7H6Cl2O
Formula Weight 177.03g/mol
MDL Number MFCD00004609
Beilstein 1938356
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
RTECS Number DO0900000