Learn More
Alfa Aesar™ 2,6-Dichlorobenzoyl chloride, 98%
Manufacturer: Alfa Aesar™ A1483909
Description
Applications2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.
Solubility
Miscible with alcohol, ether and acetone. Slightly miscible with heptane. Immiscible with water.
Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents and strong bases.
Specifications
2,6-Dichlorobenzoyl chloride | |
130°C to 132°C (15mmHg) | |
10g | |
C7H3Cl3O | |
UN3265 | |
benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043 | |
C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl | |
209.45 | |
209.46 |
>110°C (230°F) | |
15°C to 18°C | |
4659-45-4 | |
MFCD00000662 | |
639531 | |
JBLIDPPHFGWTKU-UHFFFAOYSA-N | |
2,6-dichlorobenzoyl chloride | |
78392 | |
98% |
Chemical Identifiers
4659-45-4 | |
209.45 | |
JBLIDPPHFGWTKU-UHFFFAOYSA-N | |
78392 | |
C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl |
C7H3Cl3O | |
MFCD00000662 | |
benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043 | |
2,6-dichlorobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.