Learn More
2,6-Dichlorobenzoic Acid, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113201000
Specifications
2, 6-Dichlorobenzoic acid | |
Glass bottle | |
141.0°C to 145.0°C | |
98+% | |
98.0 | |
98% min. (HPLC) | |
Cl2C6H3CO2H | |
09, 343 | |
MRUDNSFOFOQZDA-UHFFFAOYSA-N | |
2,6-dichlorobenzoic acid | |
5758 | |
191.01 | |
98+% |
Authentic | |
White to Beige | |
100g | |
50-30-6 | |
100.0 | |
C7H4Cl2O2 | |
MFCD00002418 | |
2,6 dichlorobenzoic acid, 2,6-dichloro-benzoic acid, 2,6-dichloro-benzoicacid, 2,6-dichlorobenzoicacid, 2.6-dichlorobenzoic acid, acmc-1auop, benzoic acid, 2,6-dichloro, ccris 8610, pubchem20028, unii-634ri764qt | |
C1=CC(=C(C(=C1)Cl)C(=O)O)Cl | |
191.01 | |
CHEBI:48623 | |
Fine Crystalline Powder |
Chemical Identifiers
50-30-6 | |
191.01 | |
MRUDNSFOFOQZDA-UHFFFAOYSA-N | |
5758 | |
2,6-dichlorobenzoic acid |
C7H4Cl2O2 | |
MFCD00002418 | |
2,6 dichlorobenzoic acid, 2,6-dichloro-benzoic acid, 2,6-dichloro-benzoicacid, 2,6-dichlorobenzoicacid, 2.6-dichlorobenzoic acid, acmc-1auop, benzoic acid, 2,6-dichloro, ccris 8610, pubchem20028, unii-634ri764qt | |
CHEBI:48623 | |
C1=CC(=C(C(=C1)Cl)C(=O)O)Cl |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-025-9
RTECSNumber : DG7000000
TSCA : TSCA