missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorobenzoic Acid, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113200250

 View more versions of this product

Catalog No. AC113200250



Description

Description

Specifications

Specifications

2, 6-Dichlorobenzoic acid
Glass bottle
141.0°C to 145.0°C
98+%
98.0
98% min. (HPLC)
Cl2C6H3CO2H
09, 343
MRUDNSFOFOQZDA-UHFFFAOYSA-N
2,6-dichlorobenzoic acid
5758
191.01
98+%
Authentic
White to Beige
25g
50-30-6
100.0
C7H4Cl2O2
MFCD00002418
2,6 dichlorobenzoic acid, 2,6-dichloro-benzoic acid, 2,6-dichloro-benzoicacid, 2,6-dichlorobenzoicacid, 2.6-dichlorobenzoic acid, acmc-1auop, benzoic acid, 2,6-dichloro, ccris 8610, pubchem20028, unii-634ri764qt
C1=CC(=C(C(=C1)Cl)C(=O)O)Cl
191.01
CHEBI:48623
Fine Crystalline Powder

Chemical Identifiers

50-30-6
191.01
MRUDNSFOFOQZDA-UHFFFAOYSA-N
5758
2,6-dichlorobenzoic acid
C7H4Cl2O2
MFCD00002418
2,6 dichlorobenzoic acid, 2,6-dichloro-benzoic acid, 2,6-dichloro-benzoicacid, 2,6-dichlorobenzoicacid, 2.6-dichlorobenzoic acid, acmc-1auop, benzoic acid, 2,6-dichloro, ccris 8610, pubchem20028, unii-634ri764qt
CHEBI:48623
C1=CC(=C(C(=C1)Cl)C(=O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-025-9

RTECSNumber : DG7000000

TSCA : TSCA

SDS Documents