Please login to your online account to display your discounted pricing

2,6-Dichlorobenzene-1-sulfonyl chloride, ≥95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00344EA

 View more versions of this product

Catalog No. TL00344EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 245.51
Quantity 10g
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3SO2Cl
CAS 6579-54-0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD00052311
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material 2, 6-Dichlorobenzene-1-sulfonyl chloride
Molecular Formula C6H3Cl3O2S

Specifications

Chemical Name or Material 2, 6-Dichlorobenzene-1-sulfonyl chloride
CAS 6579-54-0
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H3Cl3O2S
Linear Formula Cl2C6H3SO2Cl
Formula Weight 245.51
MDL Number MFCD00052311
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.