missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorobenzamide, 97%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 161691000

 View more versions of this product

Catalog No. AC161691000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dichlorobenzamide
Glass bottle
196.0°C to 201.0°C
97%
96.0
96% min. (GC)
Cl2C6H3CONH2
09, I, 141
JHSPCUHPSIUQRB-UHFFFAOYSA-N
2,6-dichlorobenzamide
16183
190.03
97%
Authentic
Brown-Gray to White
100g
2008-58-4
100.0
C7H5Cl2NO
MFCD00007975
2,6-bam
C1=CC(=C(C(=C1)Cl)C(=O)N)Cl
190.03
CHEBI:28435
Crystalline Powder

Chemical Identifiers

2008-58-4
190.03
JHSPCUHPSIUQRB-UHFFFAOYSA-N
16183
2,6-dichlorobenzamide
C7H5Cl2NO
MFCD00007975
2,6-bam
CHEBI:28435
C1=CC(=C(C(=C1)Cl)C(=O)N)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 217-918-4

TSCA : TSCA

SDS Documents