missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorobenzamide 97%, ACROS Organics™

Manufacturer:  Acros Organics 161690250

 View more versions of this product

Catalog No. AC161690250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dichlorobenzamide
196.0°C to 201.0°C
97%
96.0
Authentic
97%
C7H5Cl2NO
MFCD00007975
JHSPCUHPSIUQRB-UHFFFAOYSA-N
2,6-dichlorobenzamide
16183
190.03
Brown-Gray to White
25g
2008-58-4
100.0
Glass bottle
97%
Cl2C6H3CONH2
09, I, 141
C1=CC(=C(C(=C1)Cl)C(=O)N)Cl
190.023
CHEBI:28435
Crystalline Powder

Chemical Identifiers

2008-58-4
190.023
JHSPCUHPSIUQRB-UHFFFAOYSA-N
CHEBI:28435
C7H5Cl2NO
MFCD00007975
16183
Safety and Handling

Safety and Handling

EINECSNumber : 217-918-4

TSCA : TSCA

SDS Documents