missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dichlorobenzaldoxime, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1135106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dichlorobenzaldoxime | |
147°C to 151°C | |
MFCD00013938 | |
2046662 | |
YBSXDWIAUZOFFV-ONNFQVAWSA-N | |
(NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine | |
9581041 | |
97% |
25185-95-9 | |
C7H5Cl2NO | |
5 g | |
2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime | |
O\N=C\C1=C(Cl)C=CC=C1Cl | |
190.02 | |
190.03 |
Chemical Identifiers
25185-95-9 | |
190.02 | |
YBSXDWIAUZOFFV-ONNFQVAWSA-N | |
9581041 | |
O\N=C\C1=C(Cl)C=CC=C1Cl |
C7H5Cl2NO | |
MFCD00013938 | |
2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime | |
(NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-720-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only