Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloroanisole, 99%

$56.04 - $200.11

Chemical Identifiers

Chemical Name or Material 2,6-Dichloroanisole
CAS 1984-65-2
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00000575
Synonym 1,3-Dichloro-2-methoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2530209

  SDS
alfa aesar™
B2530209
10g Each for $56.04

AAB2530218

  SDS
alfa aesar™
B2530218
50g Each for $200.11
Description & Specifications

Specifications

Formula Weight 177.03
Chemical Name or Material 2,6-Dichloroanisole
TSCA TSCA
Assay 99%
Molecular Formula C7H6Cl2O
MDL Number MFCD00000575
CAS 1984-65-2
Synonym 1,3-Dichloro-2-methoxybenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 91°C (196°F)
Boiling Point 117°C (20mmHg)
Melting Point 10° to 11°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.