Learn More
2,6-Dichloroaniline, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113100250
Specifications
2, 6-Dichloroaniline | |
Authentic | |
228.0°C | |
35.0°C to 39.0°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
Cl2C6H3NH2 | |
12, 626 | |
Solubility in water: 1.6g/L (20°C); 2.6g/L (40°C) | |
C1=CC(=C(C(=C1)Cl)N)Cl | |
162.02 | |
CHEBI:46630 | |
Crystalline Solid |
118°C | |
Glass bottle | |
White to Brown | |
25g | |
608-31-1 | |
100.0 | |
C6H5Cl2N | |
MFCD00007675 | |
2,5-dichlorobenzoic acid pestanal, 2,6 dichloraniline, 2,6-dichloranilin, 2,6-dichloro aniline, 2,6-dichloro-aniline, 2,6-dichloro-phenylamine, 2,6-dichlorobenzenamine, 2,6-dichlorophenylamine, benzenamine, 2,6-dichloro, clonidine impurity c | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
2,6-dichloroaniline | |
11846 | |
162.02 | |
98% |
Chemical Identifiers
608-31-1 | |
162.02 | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
11846 | |
2,6-dichloroaniline |
C6H5Cl2N | |
MFCD00007675 | |
2,5-dichlorobenzoic acid pestanal, 2,6 dichloraniline, 2,6-dichloranilin, 2,6-dichloro aniline, 2,6-dichloro-aniline, 2,6-dichloro-phenylamine, 2,6-dichlorobenzenamine, 2,6-dichlorophenylamine, benzenamine, 2,6-dichloro, clonidine impurity c | |
CHEBI:46630 | |
C1=CC(=C(C(=C1)Cl)N)Cl |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-160-5
TSCA : TSCA
Recommended Storage : Product darkens on storage