Learn More
2,6-Dichloroaniline, 98%, Thermo Scientific Chemicals
$58.28 - $154.84
Chemical Identifiers
CAS | 608-31-1 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.01 |
MDL Number | MFCD00007675 |
InChI Key | JDMFXJULNGEPOI-UHFFFAOYSA-N |
Synonym | benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline |
PubChem CID | 11846 |
ChEBI | CHEBI:46630 |
IUPAC Name | 2,6-dichloroaniline |
SMILES | NC1=C(Cl)C=CC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC113100250
|
Thermo Scientific Chemicals
113100250 |
25 g | Glass bottle |
Each for $58.28
|
|
||||
AC113101000
|
Thermo Scientific Chemicals
113101000 |
100 g | Glass bottle |
Each for $154.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
608-31-1 | |
162.01 | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
11846 | |
2,6-dichloroaniline |
C6H5Cl2N | |
MFCD00007675 | |
benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline | |
CHEBI:46630 | |
NC1=C(Cl)C=CC=C1Cl |
Specifications
608-31-1 | |
100.0 | |
White to Brown | |
118°C | |
97.5% min. (GC) | |
C6H5Cl2N | |
MFCD00007675 | |
12, 626 | |
Solubility in water: 1.6g/L (20°C); 2.6g/L (40°C) | |
NC1=C(Cl)C=CC=C1Cl | |
162.01 | |
CHEBI:46630 | |
98% | |
2, 6-Dichloroaniline, 98% |
97.5 | |
35.0°C to 39.0°C | |
228.0°C | |
Authentic | |
Glass bottle | |
Cl2C6H3NH2 | |
25 g | |
benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline | |
JDMFXJULNGEPOI-UHFFFAOYSA-N | |
2,6-dichloroaniline | |
11846 | |
162.02 | |
Crystalline Solid |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-160-5
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO – Research Use Only