missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloroaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113101000

 View more versions of this product

Catalog No. AC113101000



Description

Description

Specifications

Specifications

2, 6-Dichloroaniline
Authentic
228.0°C
35.0°C to 39.0°C
98%
97.5
97.5% min. (GC)
Cl2C6H3NH2
12, 626
Solubility in water: 1.6g/L (20°C); 2.6g/L (40°C)
NC1=C(Cl)C=CC=C1Cl
162.01
CHEBI:46630
Crystalline Solid
118°C
Glass bottle
White to Brown
100g
608-31-1
100.0
C6H5Cl2N
MFCD00007675
2,5-dichlorobenzoic acid pestanal, 2,6 dichloraniline, 2,6-dichloranilin, 2,6-dichloro aniline, 2,6-dichloro-aniline, 2,6-dichloro-phenylamine, 2,6-dichlorobenzenamine, 2,6-dichlorophenylamine, benzenamine, 2,6-dichloro, clonidine impurity c
JDMFXJULNGEPOI-UHFFFAOYSA-N
2,6-dichloroaniline
11846
162.02
98%

Chemical Identifiers

608-31-1
162.01
JDMFXJULNGEPOI-UHFFFAOYSA-N
11846
2,6-dichloroaniline
C6H5Cl2N
MFCD00007675
2,5-dichlorobenzoic acid pestanal, 2,6 dichloraniline, 2,6-dichloranilin, 2,6-dichloro aniline, 2,6-dichloro-aniline, 2,6-dichloro-phenylamine, 2,6-dichlorobenzenamine, 2,6-dichlorophenylamine, benzenamine, 2,6-dichloro, clonidine impurity c
CHEBI:46630
NC1=C(Cl)C=CC=C1Cl
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-160-5

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS Documents
Product Certifications