Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',6'-Dichloroacetanilide, 98+%

$24.18 - $389.43

Chemical Identifiers

Chemical Name or Material 2',6'-Dichloroacetanilide
CAS 17700-54-8
Molecular Formula C8H7Cl2NO
Formula Weight 204.06
MDL Number MFCD00013629
Beilstein 2096165
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1519509

 
alfa aesar™
A1519509
10g Each for $24.18

AAA1519518

 
alfa aesar™
A1519518
50g Each for $90.96

AAA1519530

 
alfa aesar™
A1519530
250g Each for $389.43
Description & Specifications

Specifications

Formula Weight 204.06
Chemical Name or Material 2',6'-Dichloroacetanilide
Assay ≥98%
Molecular Formula C8H7Cl2NO
MDL Number MFCD00013629
CAS 17700-54-8
Beilstein 2096165
Melting Point 180° to 181°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AE1970000
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.