missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-beta-nitrostyrene, 97%

Manufacturer:  Alfa Aesar™ A1011003

 View more versions of this product

Catalog No. AAA1011003



Specifications

Specifications

2,6-Dichloro-beta-nitrostyrene
1g
C8H5Cl2NO2
1960934
VXNHQIKJDIOBEC-SNAWJCMRSA-N
1,3-dichloro-2-[(E)-2-nitroethenyl]benzene
5369223
97%
64°C to 65°C
22482-43-5
MFCD00024821
1,3-dichloro-2-2-nitrovinyl benzene, 2,6-dichloro-beta-nitrostyrene, 1,3-dichloro-2-2-nitroethenyl benzene, 1-2,6-dichlorophenyl-2-nitroethylene, 1,3-dichloro-2-e-2-nitroethenyl benzene, styrene, 2,6-dichloro-beta-nitro, 1-3,4-methylenedioxy phenyl-2-nitroethene, benzene, 1,3-dichloro-2-2-nitroethenyl, benzene,1,3-dichloro-2-2-nitroethenyl, 2-1e-2-nitrovinyl-1,3-dichlorobenzene
C1=CC(=C(C(=C1)Cl)C=C[N+](=O)[O-])Cl
218.033
218.04

Chemical Identifiers

22482-43-5
218.033
VXNHQIKJDIOBEC-SNAWJCMRSA-N
5369223
C1=CC(=C(C(=C1)Cl)C=C[N+](=O)[O-])Cl
C8H5Cl2NO2
MFCD00024821
1,3-dichloro-2-2-nitrovinyl benzene, 2,6-dichloro-beta-nitrostyrene, 1,3-dichloro-2-2-nitroethenyl benzene, 1-2,6-dichlorophenyl-2-nitroethylene, 1,3-dichloro-2-e-2-nitroethenyl benzene, styrene, 2,6-dichloro-beta-nitro, 1-3,4-methylenedioxy phenyl-2-nitroethene, benzene, 1,3-dichloro-2-2-nitroethenyl, benzene,1,3-dichloro-2-2-nitroethenyl, 2-1e-2-nitrovinyl-1,3-dichlorobenzene
1,3-dichloro-2-[(E)-2-nitroethenyl]benzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents