Learn More
2,6-Dichloro-7-deazapurine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6422703
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in DMSO, ethyl acetate and methanol. Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2,6-Dichloro-7-deazapurine | |
247°C to 250°C | |
MFCD08059278 | |
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l | |
GHXBPCSSQOKKGB-UHFFFAOYSA-N | |
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine | |
14116871 | |
≥98% |
90213-66-4 | |
C6H3Cl2N3 | |
1 g | |
Soluble in DMSO,ethyl acetate and methanol. Slightly soluble in water. | |
C1=CNC2=C1C(=NC(=N2)Cl)Cl | |
188.011 | |
188.01 | |
Solid |
Chemical Identifiers
90213-66-4 | |
188.011 | |
GHXBPCSSQOKKGB-UHFFFAOYSA-N | |
14116871 | |
C1=CNC2=C1C(=NC(=N2)Cl)Cl |
C6H3Cl2N3 | |
MFCD08059278 | |
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l | |
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only