missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-7-deazapurine, ≥98%

Manufacturer:  Alfa Aesar™ H6422706

 View more versions of this product

Catalog No. AAH6422706



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in DMSO, ethyl acetate and methanol. Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2,6-Dichloro-7-deazapurine
5g
C6H3Cl2N3
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
C1=CNC2=C1C(=NC(=N2)Cl)Cl
188.011
188.01
≥98%
247°C to 250°C
90213-66-4
MFCD08059278
GHXBPCSSQOKKGB-UHFFFAOYSA-N
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
14116871
Solid

Chemical Identifiers

90213-66-4
188.011
GHXBPCSSQOKKGB-UHFFFAOYSA-N
14116871
C1=CNC2=C1C(=NC(=N2)Cl)Cl
C6H3Cl2N3
MFCD08059278
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents