Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-5-fluoronicotinic acid, 96%

$56.20 - $194.38

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-5-fluoronicotinic acid
CAS 82671-06-5
Molecular Formula C6H2Cl2FNO2
Formula Weight 209.99
MDL Number MFCD00799517
Beilstein 4311035
Synonym 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1945414

  SDS
alfa aesar™
L1945414
25g Each for $194.38

AAL1945406

  SDS
alfa aesar™
L1945406
5g Each for $56.20
Description & Specifications

Specifications

Chemical Name or Material 2,6-Dichloro-5-fluoronicotinic acid
Quantity 25g
CAS 82671-06-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Molecular Formula C6H2Cl2FNO2
Formula Weight 209.99
MDL Number MFCD00799517
Beilstein 4311035
Melting Point 152° to 155°C
Synonym 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.