missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 338890250

 View more versions of this product

Catalog No. AC338890250


 

Specifications Description & Specifications

Specifications

2, 6-Dichloro-5-fluoro-3-pyridinecarbonitrile
85° to 92°C
82671-02-1
Glass bottle
≥94% (GC)
C6HCl2FN2
Light yellow
25g
Authentic
Crystalline Powder
95%
190.99

Chemical Identifiers

95%
190.99
≥94% (GC)
C6HCl2FN2
2, 6-Dichloro-5-fluoro-3-pyridinecarbonitrile