missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile, 95%, ACROS Organics™

$208.20 - $208.20

Chemical Identifiers

CAS 82671-02-1
Molecular Formula C6HCl2FN2
Molecular Weight (g/mol) 190.99
InChI Key DEDKKOOGYIMMBC-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-cyano-5-fluoropyridine
PubChem CID 2733849
IUPAC Name 2,6-dichloro-5-fluoropyridine-3-carbonitrile
SMILES C1=C(C(=NC(=C1F)Cl)Cl)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC338890050 SDS View Documents Thermo Fisher Scientific
338890050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC338890250 SDS View Documents Thermo Fisher Scientific
338890250
25g Glass bottle Each for $208.20
 
AC338891000 SDS View Documents Thermo Fisher Scientific
AC338891000
100g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

82671-02-1
190.99
2,6-dichloro-3-cyano-5-fluoropyridine
2,6-dichloro-5-fluoropyridine-3-carbonitrile
C6HCl2FN2
DEDKKOOGYIMMBC-UHFFFAOYSA-N
2733849
C1=C(C(=NC(=C1F)Cl)Cl)C#N

Specifications

Authentic
Yellow
5g
94% min. (GC)
2,6-dichloro-3-cyano-5-fluoropyridine
C1=C(C(=NC(=C1F)Cl)Cl)C#N
190.99
190.99
95%
Glass bottle
85.0°C to 92.0°C
82671-02-1
C6HCl2FN2
DEDKKOOGYIMMBC-UHFFFAOYSA-N
2,6-dichloro-5-fluoropyridine-3-carbonitrile
2733849
Crystalline Powder
2, 6-Dichloro-5-fluoro-3-pyridinecarbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning