Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine, 97%

$83.77 - $83.77

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine
CAS 86398-94-9
Molecular Formula C7H5Cl2F3N2
Formula Weight 245.03
MDL Number MFCD00085001
Beilstein 8319619
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2001303

  SDS
alfa aesar™
B2001303
1g Each for $83.77
Description & Specifications

Specifications

Formula Weight 245.03
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine
Assay 97%
Molecular Formula C7H5Cl2F3N2
MDL Number MFCD00085001
CAS 86398-94-9
Beilstein 8319619
Melting Point 57° to 61°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.