Please login to your online account to display your discounted pricing

2,6-Dichloro-4-(trifluoromethyl)benzonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)benzonitrile
CAS 157021-61-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H2Cl2F3N
Formula Weight 240.01
MDL Number MFCD00220879
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW01637CB

  SDS
fisher scientific company
SEW01637CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.01
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)benzonitrile
Assay Percent Range 97%
MDL Number MFCD00220879
CAS 157021-61-9
Packaging Amber Glass Bottle
Molecular Formula C8H2Cl2F3N
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.