Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)aniline
CAS 24279-39-8
Molecular Formula C7H4Cl2F3N
Formula Weight 230.02
MDL Number MFCD00052918
UN Number UN3077
Beilstein 2838819
Synonym 4-Amino-3,5-dichlorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2072803

  SDS
alfa aesar™
B2072803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2072806

  SDS
alfa aesar™
B2072806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.02
Molecular Formula C7H4Cl2F3N
Health Hazard 2 GHS H Statement
H302-H332-H315-H317
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Quantity 1g
Boiling Point 60° to 62°C (1mmHg)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
UN Number UN3077
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P362-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2838819
Flash Point 88°C (190°F)
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)aniline
MDL Number MFCD00052918
Health Hazard 1 Warning
Melting Point 34° to 37°C
Synonym 4-Amino-3,5-dichlorobenzotrifluoride
Assay 97%
CAS 24279-39-8
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P362-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332-H315-H317
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.