Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-(trifluoromethoxy)benzoic acid, 97%

$119.15 - $487.95

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)benzoic acid
CAS 886502-90-5
Molecular Formula C8H3Cl2F3O3
Formula Weight 275.01
MDL Number MFCD06660297
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2686703

  SDS
alfa aesar™
H2686703
1g Each for $119.15

AAH2686706

  SDS
alfa aesar™
H2686706
5g Each for $487.95
Description & Specifications

Specifications

Formula Weight 275.01
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)benzoic acid
Assay 97%
Molecular Formula C8H3Cl2F3O3
MDL Number MFCD06660297
CAS 886502-90-5
Melting Point 151° to 153°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.