Please login to your online account to display your discounted pricing

2,6-Dichloro-4-(trifluoromethoxy)aniline, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethoxy)aniline
CAS 99479-66-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C7H4Cl2F3NO
Formula Weight 246.02
Synonym 2-Amino-1, 3-dichloro-5-(trifluoromethoxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442300010

 
thermo fisher scientific
AC442300010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442300050

 
thermo fisher scientific
AC442300050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder, Low Melting Solid or Crystalline Needles
Assay ≥95.0%
Formula Weight 246.02
Quantity 1g
Color Colorless to white to pale brown
Melting Point 30° to 35°C
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethoxy)aniline
Assay Percent Range 96%
Molecular Formula C7H4Cl2F3NO
CAS 99479-66-0
Synonym 2-Amino-1, 3-dichloro-5-(trifluoromethoxy)benzene
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.