Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-(trifluoromethoxy)aniline, 95%

$48.95 - $48.95

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)aniline
CAS 99479-66-0
Molecular Formula C7H4Cl2F3NO
Formula Weight 246.02
MDL Number MFCD00190128
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2448204

 
alfa aesar™
B2448204
2g Each for $48.95
Description & Specifications

Specifications

Formula Weight 246.02
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)aniline
Assay 95%
Molecular Formula C7H4Cl2F3NO
MDL Number MFCD00190128
CAS 99479-66-0
Melting Point 31° to 33°C
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.