Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-nitropyridine N-oxide, 97%

$67.73 - $67.73

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-nitropyridine N-oxide
CAS 2587-01-1
Molecular Formula C5H2Cl2N2O3
Formula Weight 208.99
MDL Number MFCD00955611
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2697906

  SDS
alfa aesar™
H2697906
5g Each for $67.73
Description & Specifications

Specifications

Formula Weight 208.99
Chemical Name or Material 2,6-Dichloro-4-nitropyridine N-oxide
Assay 97%
Molecular Formula C5H2Cl2N2O3
MDL Number MFCD00955611
CAS 2587-01-1
Melting Point 176° to 180°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.