Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-nitropyridine, 97%

$48.97 - $156.70

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-nitropyridine
CAS 25194-01-8
Molecular Formula C5H2Cl2N2O2
Formula Weight 192.99
MDL Number MFCD05670545
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2713103

  SDS
alfa aesar™
H2713103
1g Each for $48.97

AAH2713106

  SDS
alfa aesar™
H2713106
5g Each for $156.70
Description & Specifications

Specifications

Formula Weight 192.99
Assay 97%
Melting Point 94° to 98°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H2Cl2N2O2
Health Hazard 1 Danger
MDL Number MFCD05670545
CAS 25194-01-8
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2,6-Dichloro-4-nitropyridine
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.