Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-nitrophenol, 98%

$72.18 - $72.18

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-nitrophenol
CAS 618-80-4
Molecular Formula C6H3Cl2NO3
Formula Weight 208
MDL Number MFCD00014715
UN Number UN2811
Beilstein 1245045
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1098814

  SDS
alfa aesar™
A1098814
25g Each for $72.18

AAA1098836

  SDS
alfa aesar™
A1098836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈124°C (dec.)
Health Hazard 1 Danger
Chemical Name or Material 2,6-Dichloro-4-nitrophenol
TSCA TSCA
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 25g
UN Number UN2811
CAS 618-80-4
Molecular Formula C6H3Cl2NO3
MDL Number MFCD00014715
Beilstein 1245045
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.