Please login to your online account to display your discounted pricing

2,6-Dichloro-4-nitroaniline 95%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113495000

 View more versions of this product

Catalog No. AC113495000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Granular Powder and Chunks
Assay ≥94%
Formula Weight 207.02
Quantity 500g
Color Yellow
EINECS Number 202-746-4
TSCA TSCA
RTECS Number BX2975000
Melting Point 187° to 192°C
Beilstein 12, 735
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2, 6-Dichloro-4-nitroaniline
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Linear Formula Cl2C6H2(NO2)NH2
MDL Number MFCD00007677
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6 H4 Cl2 N2 O2
CAS 99-30-9

Specifications

Chemical Name or Material 2, 6-Dichloro-4-nitroaniline
Name Note 95%
CAS 99-30-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6 H4 Cl2 N2 O2
Linear Formula Cl2C6H2(NO2)NH2
Formula Weight 207.02
MDL Number MFCD00007677
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents