Please login to your online account to display your discounted pricing

2,6-Dichloro-4-nitroaniline 95%, ACROS Organics™

$99.64 - $99.64

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloro-4-nitroaniline
Name Note 95%
CAS 99-30-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6 H4 Cl2 N2 O2
Linear Formula Cl2C6H2(NO2)NH2
Formula Weight 207.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113495000

  SDS
thermo fisher scientific
113495000
Glass bottle 500g Each for $99.64
Description & Specifications

Specifications

Assay ≥94%
Physical Form Granular Powder and Chunks
Formula Weight 207.02
Quantity 500g
Color Yellow
TSCA TSCA
RTECS Number BX2975000
EINECS Number 202-746-4
Melting Point 187° to 192°C
Beilstein 12, 735
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula Cl2C6H2(NO2)NH2
Name Note 95%
Chemical Name or Material 2, 6-Dichloro-4-nitroaniline
Assay Percent Range 95%
MDL Number MFCD00007677
Molecular Formula C6 H4 Cl2 N2 O2
CAS 99-30-9
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.