missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-nitroaniline, 95%

$32.01 - $99.91

Chemical Identifiers

CAS 99-30-9
Molecular Formula C6H4Cl2N2O2
Formula Weight 207.02
MDL Number MFCD00007677
Beilstein 1459581
Chemical Name or Material 2,6-Dichloro-4-nitroaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2428314 SDS Alfa Aesar™
B2428314
25g Each for $32.01
AAB2428322 SDS Alfa Aesar™
B2428322
100g Each for $33.55
AAB2428336 SDS Alfa Aesar™
B2428336
500g Each for $99.91
Specifications

Description

Applications
It is used as dyestuff, agricultural and pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

99-30-9
207.02
1459581
C6H4Cl2N2O2
MFCD00007677
2,6-Dichloro-4-nitroaniline

Specifications

25g
Recognizable
207.02
130°C (2mmHg)
185° to 192°C
95%
99-30-9
C6H4Cl2N2O2
MFCD00007677
1459581
2,6-Dichloro-4-nitroaniline

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P262-P305+P351+P338
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : BX2975000

TSCA : TSCA