missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-methylpyridine, 95%

Manufacturer:  Alfa Aesar™ H61767MD

 View more versions of this product

Catalog No. AAH61767MD



Specifications

Specifications

2,6-Dichloro-4-methylpyridine
250mg
C6H5Cl2N
2,6-bis chloranyl-4-methyl-pyridine
CC1=CC(=NC(=C1)Cl)Cl
162.013
162.02
95%
White
39621-00-6
MFCD09264302
GTHRJKYVJZJPCF-UHFFFAOYSA-N
2,6-dichloro-4-methylpyridine
10898997
Solid

Chemical Identifiers

39621-00-6
162.013
GTHRJKYVJZJPCF-UHFFFAOYSA-N
10898997
CC1=CC(=NC(=C1)Cl)Cl
C6H5Cl2N
MFCD09264302
2,6-bis chloranyl-4-methyl-pyridine
2,6-dichloro-4-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C

Documents