Learn More
2,6-Dichloro-4-isocyanatopyridine, 90%, Thermo Scientific™
Chemical Identifiers
CAS | 159178-03-7 |
---|---|
Molecular Formula | C6H2Cl2N2O |
Molecular Weight (g/mol) | 189 |
InChI Key | OCDFIQXUCLQQHY-UHFFFAOYSA-N |
Synonym | 2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci |
PubChem CID | 2781658 |
IUPAC Name | 2,6-dichloro-4-isocyanatopyridine |
SMILES | C1=C(C=C(N=C1Cl)Cl)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC451530010
|
Thermo Scientific Chemicals
451530010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC451530100
|
Thermo Scientific Chemicals
451530100 |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
159178-03-7 | |
189 | |
2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci | |
2,6-dichloro-4-isocyanatopyridine |
C6H2Cl2N2O | |
OCDFIQXUCLQQHY-UHFFFAOYSA-N | |
2781658 | |
C1=C(C=C(N=C1Cl)Cl)N=C=O |
Specifications
159178-03-7 | |
89% min. (GC) | |
C6H2Cl2N2O | |
2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci | |
C1=C(C=C(N=C1Cl)Cl)N=C=O | |
189 | |
189 | |
2,6-Dichloro-4-isocyanatopyridine |
Authentic | |
Glass Bottle | |
1g | |
OCDFIQXUCLQQHY-UHFFFAOYSA-N | |
2,6-dichloro-4-isocyanatopyridine | |
2781658 | |
90% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause allergy or asthma sympto
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
Danger
RUO â Research Use Only