missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-hydroxybenzaldehyde, 98%

$88.36 - $250.91

Chemical Identifiers

CAS 60964-09-2
Molecular Formula C7H4Cl2O2
Formula Weight 191.01
MDL Number MFCD06797919
Chemical Name or Material 2,6-Dichloro-4-hydroxybenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2839503 SDS Alfa Aesar™
H2839503
1g Each for $88.36
AAH2839506 SDS Alfa Aesar™
H2839506
5g Each for $250.91
Specifications

Description

Applications
As starting reagents proposed to use 4-hydroxy-2,6-dimethylbenzaldehyde and 2,6-dichloro-4-hydroxybenzaldehyde easily available by straightforward syntheses from commercial products.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

60964-09-2
191.01
2,6-Dichloro-4-hydroxybenzaldehyde
C7H4Cl2O2
MFCD06797919

Specifications

1g
C7H4Cl2O2
MFCD06797919
2,6-Dichloro-4-hydroxybenzaldehyde
60964-09-2
191.01
224° to 229°C
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning