Learn More
2,6-Dichloro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2839506
Description
As starting reagents proposed to use 4-hydroxy-2,6-dimethylbenzaldehyde and 2,6-dichloro-4-hydroxybenzaldehyde easily available by straightforward syntheses from commercial products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAs starting reagents proposed to use 4-hydroxy-2,6-dimethylbenzaldehyde and 2,6-dichloro-4-hydroxybenzaldehyde easily available by straightforward syntheses from commercial products.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2,6-Dichloro-4-hydroxybenzaldehyde | |
224°C to 229°C | |
MFCD06797919 | |
Air Sensitive | |
Insoluble in water. | |
OC1=CC(Cl)=C(C=O)C(Cl)=C1 | |
191.01 | |
191.01 |
60964-09-2 | |
C7H4Cl2O2 | |
5 g | |
2,6-dichloro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-dichloro-4-hydroxy, 2,6-dichloro-4-hydroxy benzaldehyde, benzaldehyde,2,6-dichloro-4-hydroxy, 2,6-bis chloranyl-4-oxidanyl-benzaldehyde | |
WWFRBIPLCLSKNH-UHFFFAOYSA-N | |
2,6-dichloro-4-hydroxybenzaldehyde | |
528382 | |
98% |
Chemical Identifiers
60964-09-2 | |
191.01 | |
WWFRBIPLCLSKNH-UHFFFAOYSA-N | |
528382 | |
OC1=CC(Cl)=C(C=O)C(Cl)=C1 |
C7H4Cl2O2 | |
MFCD06797919 | |
2,6-dichloro-4-hydroxy-benzaldehyde, benzaldehyde, 2,6-dichloro-4-hydroxy, 2,6-dichloro-4-hydroxy benzaldehyde, benzaldehyde,2,6-dichloro-4-hydroxy, 2,6-bis chloranyl-4-oxidanyl-benzaldehyde | |
2,6-dichloro-4-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only