Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-4-fluoroaniline, 98+%

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-4-fluoroaniline
CAS 344-19-4
Molecular Formula C6H4Cl2FN
Formula Weight 180.01
MDL Number MFCD00142845
UN Number UN2811
Beilstein 2718192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2544306

  SDS
alfa aesar™
B2544306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.01
Chemical Name or Material 2,6-Dichloro-4-fluoroaniline
Odor Recognizable
Assay ≥98%
Molecular Formula C6H4Cl2FN
MDL Number MFCD00142845
CAS 344-19-4
UN Number UN2811
Beilstein 2718192
Melting Point 54° to 58°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 5g

Applications
2,6-Dichloro-4-fluoroaniline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.