Please login to your online account to display your discounted pricing

2,6-Dichloro-4-(chloromethyl)pyridine, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H4Cl3N
Formula Weight 196.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00840CB

 
fisher scientific company
MO00840CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.46
Quantity 250mg
Assay 95%
CAS 101990-72-1
Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Assay Percent Range 95%
Molecular Formula C6H4Cl3N