Please login to your online account to display your discounted pricing

2,6-Dichloro-4-(chloromethyl)pyridine, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO00840CB

 View more versions of this product

Catalog No. MO00840CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.46
Quantity 250mg
Assay 95%
CAS 101990-72-1
Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Assay Percent Range 95%
Molecular Formula C6H4Cl3N

Specifications

Chemical Name or Material 2, 6-Dichloro-4-(chloromethyl)pyridine
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H4Cl3N
Formula Weight 196.46