Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-3-(trifluoromethyl)benzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzyl bromide
Molecular Formula C8H4BrCl2F3
Formula Weight 307.93
MDL Number MFCD11519349
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3179203

  SDS
alfa aesar™
H3179203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3179206

  SDS
alfa aesar™
H3179206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.93
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzyl bromide
Assay 97%
Molecular Formula C8H4BrCl2F3
MDL Number MFCD11519349
UN Number UN3261
Melting Point 69° to 72°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
2,6-Dichlorobenzyl Bromide is used in the preparation of potent epoxide hydrolyses inhibitors. Has shown thus far to relieve hypotension in lipopolysaccharide deficient murine models. And it is used in the synthesis of anti-HIV drugs based on diaryltriazine (DATA) analogues.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.