Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-3-(trifluoromethyl)benzoic acid, 97%

$73.74 - $294.00

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzoic acid
CAS 25922-41-2
Molecular Formula C8H3Cl2F3O2
Formula Weight 259.01
MDL Number MFCD09743917
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3195603

 
alfa aesar™
H3195603
1g Each for $73.74

AAH3195606

 
alfa aesar™
H3195606
5g Each for $294.00
Description & Specifications

Specifications

Formula Weight 259.01
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C8H3Cl2F3O2
MDL Number MFCD09743917
CAS 25922-41-2
Melting Point 98° to 102°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.