Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-3-(trifluoromethyl)benzaldehyde, 97%

$114.22 - $461.78

Chemical Identifiers

Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzaldehyde
CAS 134741-65-4
Molecular Formula C8H3Cl2F3O
Formula Weight 243.01
MDL Number MFCD11519344
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3246203

 
alfa aesar™
H3246203
1g Each for $114.22

AAH3246206

 
alfa aesar™
H3246206
5g Each for $461.78
Description & Specifications

Specifications

Formula Weight 243.01
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C8H3Cl2F3O
MDL Number MFCD11519344
CAS 134741-65-4
Melting Point 48° to 50°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.